Annexin V, XFD350 Conjugate
Lieferant: AAT BIOQUEST
20090
AATB20090EA 307 EUR
AATB20090
Annexin V, XFD350 Conjugate
Tests Zelltests
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
  • High stability
  • In-house quality control
  • Multiple research applications
Bitte beachten For research use only.