Annexin V, XFD350 Conjugate
Lieferant: AAT BIOQUEST
20090
AATB20090EA
307
EUR
AATB20090
Annexin V, XFD350 Conjugate
Tests
Zelltests
XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).
- High stability
- In-house quality control
- Multiple research applications
Bitte beachten For research use only.