Annexin V, XFD594 Conjugate
Lieferant: AAT BIOQUEST
20096
AATB20096EA
307
EUR
AATB20096
Annexin V, XFD594 Conjugate
Tests
Zelltests
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- High stability
- In-house quality control
- Multiple research applications
Bitte beachten For research use only.