DABCYL-DBCO FRET-Akzeptor-Vorläufer

Lieferant: AAT BIOQUEST
Achtung

2010
AATB2010EA 439 EUR
AATB2010
DABCYL-DBCO FRET-Akzeptor-Vorläufer
DABCYL-DBCO

DABCYL is one of the most common dark quenchers used to prepare a variety of FRET-based nucleic acid probes and protease substrates.


  • FRET probe precursor

Molecular Weight: 527,63 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d30e46063facd63a41623a332e8d18c73092c72c&vid=aadbcb70ffe03c3adab706a1911ae0c38eb923db&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR