6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE) einfaches Isomer ≥90%
Lieferant: AAT BIOQUEST
Synonyme:
6-HEX, NHS ester
202
AATB202EA
230
EUR
AATB202
6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE) einfaches Isomer ≥90%
6-Carboxy-2',4,4',5',7,7'-hexachlorofluorescein succinimidyl ester (6-HEX SE)
6-HEX, SE is the amine-reactive succinimidyl ester of 6-HEX acid.
- Useful for Genetic Research
- Excellent Quality
Bitte beachten For research use only.
Formel:
C₂₅H₉Cl₆NO₉ Molecular Weight: 680,06 g/mol |
CAS-Nummer:
2129651-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...