6-[(2,4-Dinitrophenyl)amino]hexansäure
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
6-((2,4-Dinitrophenyl)amino)hexanoic acid, Dnp-ε-Amino-N-Caproic Acid, DNP-X acid, N-Dnp-6-aminohexanoic Acid
2020
AATB2020EA
152
EUR
AATB2020
6-[(2,4-Dinitrophenyl)amino]hexansäure
6-[(2,4-Dinitrophenyl)amino]hexansäure
An amine-reactive building block for developing a probe that can be recognized by anti-DNP antibodies; Also an excellent amine-reactive FRET quencher paired with Trp or Tyr.
Formel:
C₁₂H₁₅N₃O₆ Molecular Weight: 297,27 g/mol |
MDL:
MFCD00038470 CAS-Nummer: 10466-72-5 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...