N-(2,4-Dinitrophenyl)-6-amino-capronsäure-N-succinimidylester ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-(2,4-Dinitrophenyl)-6-aminohexanoic Acid N-Succinimidyl Ester, DNP-X acid, SE

2021
AATB2021EA 152 EUR
AATB2021
N-(2,4-Dinitrophenyl)-6-amino-capronsäure-N-succinimidylester ≥90%
N-(2,4-Dinitrophenyl)-6-amino-capronsäure-N-succinimidylester

An amine-reactive building block for developing a probe that can be recognized by anti-DNP antibodies; Also an excellent amine-reactive FRET quencher paired with Trp or Tyr.

Formel: C₁₆H₁₈N₄O₈
Molecular Weight: 394,34 g/mol
MDL: MFCD00191630
CAS-Nummer: 82321-04-8
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR