DNP-PEG4-acid FRET-Quencher-Vorläufer
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
1-((2,4-Dinitrophenyl)amino)-3,6,9,12-tetraoxapentadecan-15-oic acid
2022
AATB2022EA
137
EUR
AATB2022
DNP-PEG4-acid FRET-Quencher-Vorläufer
DNP-PEG4-acid
Compared to the commonly used DNP-X acid (#2020), DNP-PEG4 acid has much better water solubility.
- FRET probe precursor
Formel:
C₁₇H₂₅N₃O₁₀ |
MDL:
MFCD13184986 CAS-Nummer: 858126-76-8 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...