DNP-PEG4-NHS ester FRET-Quencher-Vorläufer

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2,5-Dioxopyrrolidin-1-yl 1-((2,4-dinitrophenyl)amino)-3,6,9,12-tetraoxapentadecan-15-oate

2023
AATB2023EA 137 EUR
AATB2023
DNP-PEG4-NHS ester FRET-Quencher-Vorläufer
DNP-PEG4-NHS ester

Compared to the commonly used DNP-X SE (#2021), DNP-PEG4 NHS ester has much better water solubility.


  • FRET probe precursor

Formel: C₂₁H₂₈N₄O₁₂
Molecular Weight: 528,467 g/mol
MDL: MFCD13184987
CAS-Nummer: 858126-78-0
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62a674a9619fd8d3d62d645af6ad61a9762e7eb5&vid=62a5082235ab7e1c785c572a771b33912f67aeeb&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR