6-Carboxy-4',5'-dichloro-2',7'-dimethoxyfluorescein, succinimidyl ester ≥90%

Lieferant: AAT BIOQUEST
Achtung

203
AATB203EA 307 EUR
AATB203
6-Carboxy-4',5'-dichloro-2',7'-dimethoxyfluorescein, succinimidyl ester ≥90%
6-Carboxy-4',5'-dichloro-2',7'-dimethoxyfluorescein, succinimidyl ester

6-JOE has similar absorption and emission spectra to those of rhodamine 6G, and its SE is predominantly used in automated DNA sequencing.

Formel: C₂₇H₁₇Cl₂NO₁₁
Molecular Weight: 602,34 g/mol
CAS-Nummer: 113394-23-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR