Native 2',3'-cGAMP (from Mammalian cells), Biotin
Lieferant: AAT BIOQUEST
20316
AATB20316EA 1370 EUR
AATB20316
Native 2',3'-cGAMP (from Mammalian cells), Biotin
Proteine und Peptide
2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate 'adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.
  • Essential component of intracellular signaling
Bitte beachten For research use only.