Native 2',3'-cGAMP (from Mammalian cells), Cy5®
Lieferant: AAT BIOQUEST
20318
AATB20318EA 1370 EUR
AATB20318
Native 2',3'-cGAMP (from Mammalian cells), Cy5®
Proteine und Peptide
2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate"adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.
  • Essential component of intracellular signaling
Bitte beachten For research use only.