2’,3’-cGAMP-iFluor 647 Konjugat
Lieferant: AAT BIOQUEST
20322
AATB20322EA
1330
EUR
AATB20322
2’,3’-cGAMP-iFluor 647 Konjugat
2’,3’-cGAMP-iFluor 647
iFluor® 647-labeled cGAMP provides an excellent fluorescent probe for developing fluorescence-based assays and tests for detecting cGAMP or monitoring its activity.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only