QXY21 NHS ester [equivalent to QSY-21 NHS ester]
Lieferant: AAT BIOQUEST
2033
AATB2033EA 309 EUR
AATB2033
QXY21 NHS ester [equivalent to QSY-21 NHS ester]
QXY21 NHS ester
QXY21 is the same molecule to QSY-21 acid (ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only