2’,3’-cGAMP-DBCO Konjugat
Lieferant: AAT BIOQUEST
20330
AATB20330EA
1330
EUR
AATB20330
2’,3’-cGAMP-DBCO Konjugat
2’,3’-cGAMP-DBCO
DBCO (dibenzocyclooctyne) click chemistry is a bioconjugation technique used to covalently attach molecules to biological targets, such as proteins, peptides, and nucleic acids.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only