2’,3’-cGAMP azide
Lieferant: AAT Bioquest
2’,3’-cGAMP azide is an excellent reagent to conjugate 2’,3’-cGAMP to alkyne-modified biological targets, such as proteins, peptides, and nucleic acids.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only