2’,3’-cGAMP succinimidyl ester
Lieferant: AAT BIOQUEST
20334
AATB20334EA 1330 EUR
AATB20334
2’,3’-cGAMP succinimidyl ester
2’,3’-cGAMP succinimidyl ester
2’,3’-cGAMP succinimidyl ester is an excellent reagent to covalently conjugate 2’,3’-cGAMP to biological targets, such as proteins, peptides, and nucleic acids.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only