Furaptra (Mag-Fura-2) tetrapotassium salt Magnesium-Indikator
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Mag-Fura-2 tetrapotassium salt
20383
AATB20383EA
307
EUR
AATB20383
Furaptra (Mag-Fura-2) tetrapotassium salt Magnesium-Indikator
Furaptra (Mag-Fura-2) tetrapotassium salt
Mag-Fura-2, AM is an intracellular magnesium indicator that is ratiometric and UV light-excitable.
- Cell-permeable
- Useful for both magnesium and high-concentration calcium detection
Formel:
C₁₈H₁₀K₄N₂O₁₁ Molecular Weight: 587 g/mol |
CAS-Nummer:
132319-57-4 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...