Mag-Fluo-4 AM
Lieferant: AAT BIOQUEST
|
Achtung
|
20401
AATB20401EA
230
EUR
AATB20401
Mag-Fluo-4 AM
Mag-Fluo-4 AM
The cell-permeant Mag-Fluo-4 AM is an analog of Fluo-4 AM with a Kd for Mg ion of 4.7 mM and a Kd for Ca ion of 22 µM, making it useful as an intracellular Mg ion indicator as well as a low-affinity Ca ion indicator.
- Suitable for either mg or ca detection
- Steady signal
- Cell permeant
Bitte beachten For research use only.
Molecular Weight:
817,65 g/mol |
CAS-Nummer:
1097709-31-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...