Mag-520™ potassium salt
Lieferant: AAT BIOQUEST
|
Achtung
|
20404
AATB20404EA
460
EUR
AATB20404
Mag-520™ potassium salt
Mag-520™ potassium salt
A vast majority of the existing magnesium ion indicators are based on tricarboxylate APTRA chelator derived from the popular tetracarboxylate BAPTA calcium ion chelator.
- Higher sensitivity
- Specific to intracellular calcium
- Steady signal
- Cell impermeant
Bitte beachten For research use only.
Molecular Weight:
511,51 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...