Cal-500™ potassium salt
Lieferant: AAT Bioquest
|
Achtung
|
20410
AATB20410EA
307
EUR
AATB20410
Cal-500™ potassium salt
Cal-500™ potassium salt
Calcium measurement is critical for numerous biological investigations.
- Higher sensitivity
- Specific to intracellular calcium
- Steady signal
- Cell impermeant
Bitte beachten For research use only.
ADR:
5,II |