Native Dextran (from Eukaryotic cells), Rhodamine 2
Lieferant: AAT BIOQUEST
20451
AATB20451EA
614
EUR
AATB20451
Native Dextran (from Eukaryotic cells), Rhodamine 2
Proteine und Peptide
Dextran-conjugated calcium indicators are stably retained within neurons.
- Well-retained in neurons
- 10000 mw
- Useful for in vivo imaging
Bitte beachten For research use only.