Cal-670™ potassium salt
Lieferant: AAT BIOQUEST
20455
AATB20455EA
619
EUR
AATB20455
Cal-670™ potassium salt
Cal-670™ potassium salt
Cal-670™ is a near infrared (NIR) calcium indicator with maximum emission at ~675 nm.
- Stable for 12 months
- Kd ~853 nM
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only