Cal-670™-Dextran Konjugat, M.W. 3000
Lieferant: AAT Bioquest
20456
AATB20456EA 775 EUR
AATB20456
Cal-670™-Dextran Konjugat, M.W. 3000
Cal-670™-Dextran
Cal-670™ is a near infrared (NIR) calcium indicator with maximum emission at ~675 nm.
  • Stable for 12 months
  • Kd ~853 nM
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only