Cal-670™-Dextran Konjugat, M.W. 3000
Lieferant: AAT Bioquest
Cal-670™ is a near infrared (NIR) calcium indicator with maximum emission at ~675 nm.
- Stable for 12 months
- Kd ~853 nM
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only