Cal-670™-Dextran Konjugat, M.W. 10000
Lieferant: AAT BIOQUEST
20457
AATB20457EA
775
EUR
AATB20457
Cal-670™-Dextran Konjugat, M.W. 10000
Cal-670™-Dextran
Cal-670™ is a near infrared (NIR) calcium indicator with maximum emission at ~675 nm.
- Stable for 12 months
- Kd ~853 nM
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only