Cal-770™ potassium salt
Lieferant: AAT BIOQUEST
|
Achtung
|
20460
AATB20460EA
614
EUR
AATB20460
Cal-770™ potassium salt
Cal-770™ potassium salt
Calcium measurement is critical for numerous biological investigations.
- Higher sensitivity
- Specific to intracellular calcium
- Steady signal
- Cell impermeant
Bitte beachten For research use only.
Molecular Weight:
1688,58 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...