6-(4,6-Dichlorotriazinyl)aminofluorescein

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 6-DTAF

205
AATB205EA 3030 EUR
AATB205
6-(4,6-Dichlorotriazinyl)aminofluorescein
6-(4,6-Dichlorotriazinyl)aminofluorescein

Compared to 5-DTAF, the 6-isomer is more preferably used for labeling oligonucleotides and nucleic acids.

Formel: C₂₃H₁₂Cl₂N₄O₅
Molecular Weight: 495,28 g/mol
CAS-Nummer: 118357-32-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR