Tide Quencher™ 2WS acid, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2WS acid

2050
AATB2050EA 307 EUR
AATB2050
Tide Quencher™ 2WS acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2WS acid

TQ2WS is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 490,6 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a73fa7fc728a211558ae9d063219f7431da5843&vid=a5a485ff19a3991cd9e72824d2607a239f700646&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR