Cal Green™ 1 hexapotassium salt
Lieferant: AAT BIOQUEST
Achtung
20500
AATB20500EA 230 EUR
AATB20500
Cal Green™ 1 hexapotassium salt
Cal Green™ 1 hexapotassium salt
Cal Green™ 1 (AAT Bioquest) is the same molecule to Calcium Green-1 (Invitrogen).
  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant
Bitte beachten For research use only.
ADR: 5,II