Cal-590™ AM
Lieferant: AAT Bioquest
|
Achtung
|
20510
20511
20512
AATB20510EA
307
EUR
AATB20510
AATB20511
AATB20512
Cal-590™ AM
Cal-590™ AM
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
Bitte beachten For research use only.
ADR:
5,II |