Cal-590™ sodium salt
Lieferant: AAT Bioquest
Achtung
20515
AATB20515EA 307 EUR
AATB20515
Cal-590™ sodium salt
Cal-590™ sodium salt
Calcium measurement is critical for numerous biological investigations.
  • Improved sensitivity
  • Improved calcium response
  • Long-wavelength emission
  • Cell impermeant
Bitte beachten For research use only.
ADR: 5,II