Cal-590L®-Dextran Konjugat, M.W. 10000
Lieferant: AAT Bioquest
20525
AATB20525EA 561 EUR
AATB20525
Cal-590L®-Dextran Konjugat, M.W. 10000
Cal-590L®-Dextran
Among the fluorescent calcium indicator dextran conjugates, Cal-590L dextran conjugates might be a better choice than other red fluorescent dextran conjugates due to its higher fluorescence quantum yield and larger fluorescence enhancement by calcium.
  • Stable for 12 months
  • Kd ~230 uM
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only