Cal-590L®/Cy5-Dextran Konjugat, M.W. 10000, RatioWorks™
Lieferant: AAT BIOQUEST
20527
AATB20527EA
702
EUR
AATB20527
Cal-590L®/Cy5-Dextran Konjugat, M.W. 10000, RatioWorks™
Cal-590L®/Cy5-Dextran
RatioWorks™ Cal-590L/Cy5-Dextran Conjugate carries a calcium-independent Cy5 fluorphore as a reference color for radiometric measurements.
- Stable for 12 months
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only