Cal-630™ sodium salt

Lieferant: AAT BIOQUEST
Achtung

20535
AATB20535EA 307 EUR
AATB20535
Cal-630™ sodium salt
Cal-630™ sodium salt

Calcium measurement is critical for numerous biological investigations.


  • Improved sensitivity
  • Improved calcium response
  • Long-wavelength emission
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 1042,45 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR