Cal-630™ potassium salt
Lieferant: AAT BIOQUEST
|
Achtung
|
20538
AATB20538EA
307
EUR
AATB20538
Cal-630™ potassium salt
Cal-630™ potassium salt
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
- Cell impermeant
Bitte beachten For research use only.
Molecular Weight:
1139,11 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...