Cal-630™-Dextran Konjugat, M.W. 10,000
Lieferant: AAT BIOQUEST
|
Achtung
|
20546
AATB20546EA
460
EUR
AATB20546
Cal-630™-Dextran Konjugat, M.W. 10,000
Cal-630™-Dextran
Calcium measurement is critical for numerous biological investigations.
- Improved sensitivity
- Improved calcium response
- Long-wavelength emission
- Cell impermeant
Bitte beachten For research use only.
Molecular Weight:
11000 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...