Fluo-5N pentapotassium Salt zellundurchlässig

Lieferant: AAT BIOQUEST
Achtung

20567
AATB20567EA 307 EUR
AATB20567
Fluo-5N pentapotassium Salt zellundurchlässig
Fluo-5N pentapotassium Salt

Fluo-5N is an analog of Fluo-4 with lower calcium-binding affinity (Kd = ~90 µM), making it suitable for detecting intracellular calcium levels in the range of 1 µM to 1 mM that would saturate the response of Fluo-4.


  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 958,06 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR