Tide Quencher™ 2WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2WS, SE

2058
AATB2058EA 307 EUR
AATB2058
Tide Quencher™ 2WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2WS succinimidyl ester

TQ2WS is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 688,86 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7924f9eb88194e31aa6efc20f98e270f04fe7dc0&vid=c21d49e88d478ca5b6ee08fee324f76fba0e3fdd&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR