Cal Red™ R525/650 potassium salt

Lieferant: AAT BIOQUEST
Achtung

20588
AATB20588EA 307 EUR
AATB20588
Cal Red™ R525/650 potassium salt
Cal Red™ R525/650 potassium salt

The intracellular calcium flux assay is a widely used method for monitoring the activities of GPCRs and calcium channels.


  • Calcium-dependent fluorescence
  • Improved sensitivity
  • Compatible with hts filters
  • Ratiometric detection
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 1000 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR