Tide Quencher™ 2WS maleimide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2WS maleimide

2059
AATB2059EA 307 EUR
AATB2059
Tide Quencher™ 2WS maleimide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2WS maleimide

TQ2WS is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 612,72 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33c2beb56726c0bec87264e7bd19b8d2a2ebc3c4&vid=c05148dce560b499507dcae6233214f22bc3b086&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR