4'-(Aminomethyl)fluorescein [AMF] ≥95%

Lieferant: AAT BIOQUEST
Achtung

206
AATB206EA 152 EUR
AATB206
4'-(Aminomethyl)fluorescein [AMF] ≥95%
4'-(Aminomethyl)fluorescein [AMF]

4'-(Aminomethyl)fluorescein is an excellent building block for developing fluoresceinated bioconjugates.

Formel: C₂₁H₁₆ClNO₅
CAS-Nummer: 91539-64-9
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR