Tide Quencher™ 4WS acid, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ4WS acid

2060
AATB2060EA 307 EUR
AATB2060
Tide Quencher™ 4WS acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4WS acid

TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 912,99 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b5ebb20f50167ebf150cc23e51aa8005414c7c1&vid=06402d1900ee430b862645aa9bea2730ab48b26b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR