Dextran, M.W. 10,000 Cal-520L™ labelled
Lieferant: AAT BIOQUEST
20602
AATB20602EA
460
EUR
AATB20602
Dextran, M.W. 10,000 Cal-520L™ labelled
Dextran
Calcium measurement is critical for numerous biological investigations.
- Well-retained in cells
- 10000 mw
- Useful for in vivo imaging
From Eukaryotic cells.
Bitte beachten For research use only.
Formel:
(C₆H₁₀O₅)n |
MDL:
MFCD00130935 CAS-Nummer: 9004-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...