Cal-520L®/Cy5-Dextran Konjugat, M.W. 10,000, RatioWorks™
Lieferant: AAT Bioquest
|
Achtung
|
20603
AATB20603EA
769
EUR
AATB20603
Cal-520L®/Cy5-Dextran Konjugat, M.W. 10,000, RatioWorks™
Cal-520L®/Cy5-Dextran
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Improved sensitivity
- Compatible with hts filters
- Ratiometric detection
- Cell impermeant
Bitte beachten For research use only.
ADR:
5,II |