Cal-520L®/Cy5-Dextran Konjugat, M.W. 10,000, RatioWorks™
Lieferant: AAT Bioquest
Achtung
20603
AATB20603EA 769 EUR
AATB20603
Cal-520L®/Cy5-Dextran Konjugat, M.W. 10,000, RatioWorks™
Cal-520L®/Cy5-Dextran
Calcium measurement is critical for numerous biological investigations.
  • Calcium-dependent fluorescence
  • Improved sensitivity
  • Compatible with hts filters
  • Ratiometric detection
  • Cell impermeant
Bitte beachten For research use only.
ADR: 5,II