Dextran, M.W. 10,000 Cal-520®/zFluor 647™ labelled, RatioWorks™
Lieferant: AAT BIOQUEST
20604
AATB20604EA
711
EUR
AATB20604
Dextran, M.W. 10,000 Cal-520®/zFluor 647™ labelled, RatioWorks™
Dextran
Calcium measurement is critical for numerous biological investigations.
- Well-retained in cells
- 10000 MW
- Useful for in vivo imaging
- Ratiometric imaging
From Eukaryotic cells.
Bitte beachten For research use only.
Formel:
(C₆H₁₀O₅)n |
MDL:
MFCD00130935 CAS-Nummer: 9004-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...