Native Biotin (from Eukaryotic cells), Cal-520®
Lieferant: AAT Bioquest
20605
AATB20605EA
460
EUR
AATB20605
Native Biotin (from Eukaryotic cells), Cal-520®
Proteine und Peptide
Biotin and biocytin bind with high affinity to both avidins and streptavidins.
- High specificity
- Sensitive detection
- Multiple applications
Bitte beachten For research use only.