Cal-520® NHS Ester

Lieferant: AAT BIOQUEST
Achtung

20609
AATB20609EA 614 EUR
AATB20609
Cal-520® NHS Ester
Cal-520® NHS Ester

Calcium measurement is critical for numerous biological investigations.


  • Calcium-dependent fluorescence
  • Steady signal
  • Improved photostability


Bitte beachten For research use only.

Molecular Weight: 1048,9 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR