Tide Quencher™ 4WS amine, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ4WS amine

2061
AATB2061EA 307 EUR
AATB2061
Tide Quencher™ 4WS amine, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4WS amine

TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 1069,1 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45c7dfb95d61d0246a82680a322c4dbb1f6df887&vid=f7903229177928d832742a895e4c588e234264f4&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR