Cal-520L™ maleimide
Lieferant: AAT BIOQUEST
Achtung
20611
AATB20611EA 614 EUR
AATB20611
Cal-520L™ maleimide
Cal-520L™ maleimide
Calcium measurement is critical for numerous biological investigations.
  • Calcium-dependent fluorescence
  • Steady signal
  • Improved photostability
Bitte beachten For research use only.
ADR: 5,II