Cal-520® amine
Lieferant: AAT BIOQUEST
20612
AATB20612EA
619
EUR
AATB20612
Cal-520® amine
Cal-520® amine
Cal-520® amine is an excellent building block that can be readily used to prepare a calcium-sensitive bioconjugate for monitoring calcium change spatially for a specific target.
- Stable for 12 months
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only