Cal-520® azide
Lieferant: AAT BIOQUEST
20613
AATB20613EA 619 EUR
AATB20613
Cal-520® azide
Cal-520® azide
Cal-520® azide is an excellent building block that can be readily used to prepare a calcium-sensitive bioconjugate for monitoring calcium change spatially for a specific target.
  • Stable for 12 months
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only